Merck
Z229776
Sigma-Aldrich
Drying tube complete for Büchi® models B-580 & B-585 ovens
borosilicate glass, includes drying tube, desiccant bulb, and removeable vacuum connections
Synonym(e):
Büchi Drying Tube, Oven Drying Tube
Größe auswählen
1 EA
CHF 903.00
CHF 903.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 903.00
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.43
CHF 903.00
Check Cart for Availability
Materialien
borosilicate glass
Verpackung
pack of 1 ea
Hersteller/Markenname
Büchi 037010
Allgemeine Beschreibung
The Buchi B-585 glass drying oven (Z563080 and Z616818) has a completely replaceable drying tube assembly. Assembly comes with the drying tube, desiccant bulb, and removeable vacuum connections. A maximum of 100 mL to 250 mL can be dried using the drying accessory. If required, the drying tube can be ventilated with protective gas. The drying hood can be filled with a drying solution (silica gel, CaCl2 or P2O5) to dry strongly hygroscopic substances.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Hi, Can I ask what is the radius of the drying tube.
    1 answer
    1. The tube has a diameter of 50 mm, with the exception of the flange, which is 56 mm. The flange ring diameter is 68 mm and including the flange coupliing, a total of 74 mm at the widest point.