Merck
Z170011
Scienceware® glove box adapter
Threaded metal
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Etikett
D
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art H50025-0010
Allgemeine Beschreibung
Tube, 21/2 in. L with 3/8 in./27 thread along complete length. Inserted in a 3/8 in. threaded hole drilled anywhere in glove box, gives threaded tube on inside and outside of box for attaching needle valves or hose ends. Complete with nuts, gaskets, and washers to seal hole.
Rechtliche Hinweise
Scienceware is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)