Merck
Z147931
Sigma-Aldrich
Glass sample holder
Boat, size 1 in. × 6 in.
Größe auswählen
1 EA
CHF 198.00
CHF 198.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 198.00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.43
CHF 198.00
Check Cart for Availability
Materialien
borosilicate glass
Parameter
230-490 °C temp. limit
Größe
1 in. × 6 in.
Allgemeine Beschreibung
These sample holders come in both boat and tube designs, made of borosilicate glass. The glass vessel is ideal for transporting and drying chemical samples in the Aldrich Abderhalden apparatus, in desiccators, or in any drying oven. It has a molded end piece to grip and easier handling of the boat.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)