Merck
Z137243
Sigma-Aldrich
Ace three-neck round-bottom flask with threaded side-arm
capacity 500 mL
Synonym(e):
bottom flask, flask round, round flask
Größe auswählen
1 EA
CHF 258.00
CHF 258.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 258.00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
CHF 258.00
Check Cart for Availability
Materialien
borosilicate glass
round bottom flask
three-neck (Vertical)
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 6954-75
Kapazität
500 mL
Allgemeine Beschreibung
Three-neck, vertical. 24/40 joints. One threaded side-arm with bushing and O-ring, except 500 mL size which has angled side necks.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)