Merck
Z116165
Sigma-Aldrich
Aldrich® large-capacity Abderhalden drying apparatus replacement parts
desiccant chamber
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
borosilicate glass
Allgemeine Beschreibung
The desiccant chamber is a replacment part for the Aldrich large capacity Abderhalden drying apparatus, Z103187.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)