Merck
TEM-LC305NI
Sigma-Aldrich
Lacey Carbon Supported Nickel Grids
size 300 mesh, size 50 nm , hole size, box of 5 ×
Synonym(e):
Lacey/Carbon, 300 Mesh, Nickel, 5pk
About This Item
UNSPSC-Code:
41111761
Verpackung
box of 5 ×
Größe
300 mesh
50 nm , hole size
Allgemeine Beschreibung
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)