Merck
TEM-LC200NICC25
Sigma-Aldrich
Continuous Ultrathin Carbon Film Coated Lacey Carbon Supported Nickel Grid
size 200 mesh, box of 25
Synonym(e):
CONTINUOUS LAYER OF ULTRATHIN CARBON FILM ON LC200NI 25/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Das Produkt TEM-LC200NICC25 ist derzeit nicht erhältlich. Setzen Sie sich mit dem technischen Dienst in Verbindung.
Verpackung
box of 25
Größe
200 mesh
Allgemeine Beschreibung
Lacey Carbon Film Coated with a Continuous Layer of Ultrathin Carbon Film. Nickel Grid, 200 mesh. The continuous ultrathin film on lacey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. The film (less than 3nm thick) lies across a carbon lacey film supported by a 200 mesh grid. The size of the holes in the lacey film differ widely from batch to batch but are generally in the range of ¼ μm to 5μm, which gives the equivalent support of at least 6000 mesh grid. Specimen material lying over the covered holes can be imaged in the TEM with practically no interference from the carbon film supporting it. This product is ideal for looking at nanotubes, virus particles and other small particulate material.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)