TEM-3752
HPM100 Leica adaptor for High Pressure Freezing with CryoCapsule
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111761
Empfohlene Produkte
Allgemeine Beschreibung
Adaptor that loads CryoCapsule, and allow High Pressure Freezing Using HPM100 machine for correlative light and electron microscopy (CLEM). For step-by-step usage please see: https://www.sciencedirect.com/science/article/pii/B9780128010754000124
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)