Merck
SYNS521000
Synthware storage flask with #15 o-ring joint and high vacuum valve
capacity 1,000 mL
Synonym(e):
Synthware S521000
Größe auswählen
1 EA
CHF 121.00
CHF 121.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 121.00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
CHF 121.00
Check Cart for Availability
Materialien
borosilicate glass flask
round bottom
single-neck
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S521000
Kapazität
1,000 mL
Allgemeine Beschreibung
Flask has a #15 O-ring joint on the sidearm and a high vacuum valve to control gas or liquid flow into the vessel. Supplied complete with one #116 Viton® O-ring.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)