Merck
SYNM202405
Synthware vacuum manifold with high vacuum valves
number of ports, 5, Hose Connection: right
Synonym(e):
Synthware M202405, vacuum manifold, vacuum unit
Größe auswählen
1 EA
CHF 686.00
CHF 686.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 686.00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
CHF 686.00
Check Cart for Availability
Leistungsmerkmale
number of ports 5
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware M202405
Allgemeine Beschreibung
With a serrated hose connection(10 mm O.D.) at right hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)