SYNF621460
Synthware™ pressure equalizing addition funnel
60 mL, joint: ST/NS 14/20, stopcock: PTFE
Synonym(e):
Synthware F621460, graduated addition funnel
Empfohlene Produkte
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F621460
Kapazität
60 mL
60 mL
Allgemeine Beschreibung
With 1:5 PTFE stopcock
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.