Merck
SYNF41611L
Synthware two neck round bottom flask with angled side neck
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Synonym(e):
Synthware F41611L, 2-neck round flask
Größe auswählen
1 EA
CHF 99.10
CHF 99.10
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 99.10
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
CHF 99.10
Check Cart for Availability
Materialien
borosilicate glass flask
round bottom
two-neck (angled side neck)
Verbindung
center joint: ST/NS24/40
side joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F41611L
Kapazität
1,000 mL
1000 mL
1000 mL
Allgemeine Beschreibung
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)