Merck
SYNE112585X
Synthware extraction thimble
O.D. 25 mm, porosity: extra coarse, H 85 mm
Synonym(e):
Synthware E112585X, glass thimble
Größe auswählen
1 EA
CHF 54.00
CHF 54.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 54.00
About This Item
UNSPSC-Code:
41104817
NACRES:
NB.51
CHF 54.00
Check Cart for Availability
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware E112585X
Höhe
85 mm
AD
25 mm
Verwandte Kategorien
Allgemeine Beschreibung
All glass extraction thimble permits viewing extraction process and subsequent weighing. C-Coarse, XC-Extra Coarse. Small thimble is used with C243445 and E103424; Medium thimble is used with C244500 and E104524; Large thimble is used with C245550 and E105550.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)