Merck
SYNC509180
Synthware Liebig condenser with 8 mm OD removable hose connections
joint: ST/NS 19/22, jacket L 180 mm
Synonym(e):
Synthware C509180, straight condenser
Größe auswählen
1 EA
CHF 102.00
CHF 102.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 102.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
CHF 102.00
Check Cart for Availability
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C509180
Mantel L
180 mm
Verwandte Kategorien
Allgemeine Beschreibung
Synthware Liebig condenser features a 180 mm jacketed length for superior cooling efficiency. With standard taper outer joint at the top (19/22) and a lower inner drip tip joint. Made from durable borosilicate glass.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)