SYNC366030
Synthware™ chromatography column with PTFE stopcock
I.D. 53.0 mm, L 305 mm
Synonym(e):
Synthware C366030, glass chromatograpy column
Empfohlene Produkte
Qualität
for chromatography
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C366030
ID
53.0 mm
Länge
305 mm
Allgemeine Beschreibung
With reinforced beaded top. All Synthware columns are heavy wall designed to increase safety.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.