Merck
SYNA921922
Synthware vacuum or inert gas adapter
male joint: ST/NS 19/22
Synonym(e):
Synthware A921922, glass vacuum adapter
Größe auswählen
1 EA
CHF 23.20
CHF 23.20
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 23.20
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
CHF 23.20
Check Cart for Availability
Verbindung
male joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A921922
Verwandte Kategorien
Allgemeine Beschreibung
Synthware vacuum or gas inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection (8 mm O.D.) is at a 90 degree angle to the standard taper inner joint (19/22).
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)