Merck
SYNA532440
Synthware inlet or thermometer adapter with hose connection
joint: ST/NS 24/40, O.D. 5.0-7.5 mm
Synonym(e):
Synthware A532440, thermometer inlet adapter
Größe auswählen
1 EA
CHF 40.40
CHF 40.40
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 40.40
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
CHF 40.40
Check Cart for Availability
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A532440
AD
5.0-7.5 mm
Allgemeine Beschreibung
Same as thermometer adapter but with 8mm or 10mm hose connection for N2/Ar introduction or vacuum. Supplied with one Nylon cap and one Viton® O-ring.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)