Merck
SLW4542/02
Pyrex® Screwcap, bottle pourer with GL 45 screwthread
Synonym(e):
Pyrex, SciLabware 4542/02
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Materialien
blue polypropylene cap
white PTFE tube
Hersteller/Markenname
4542/02
SciLabware
Fitting
GL 45
Allgemeine Beschreibung
  • Manufactured from white PTFE and blue polypropylene
  • Aids the dispensing of liquids from bottles and flasks with a GL 45 screwthread
  • Provides greater control of flow with large capacity bottles that can be difficult to handle
  • Supplied complete with venting tube and attached PTFE cap
  • Temperature resistance to 140 deg. C
  • Autoclavable
  • Dishwasher safe
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)