Merck
SLW3298/02
Pyrex® Burettes, class B, Sofnol, automatic zero, Rotaflo(R) stopcock, PTFE key
10 mL
Synonym(e):
Pyrex, SciLabware 3298/02
Größe auswählen
5 EA
CHF 519.00
CHF 519.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
5 EA
CHF 519.00
About This Item
UNSPSC-Code:
41121808
NACRES:
NB.43
CHF 519.00
Check Cart for Availability
Materialien
PTFE key
Leistungsmerkmale
graduations 0.1 mL
Hersteller/Markenname
3298/02
SciLabware
Kapazität
10 mL
Allgemeine Beschreibung
  • Complies with ISO 385
  • With PTFE Rotaflo Stopcock for grease-free performance and fine drop-wise control
  • 3mm bore PTFE key
  • Graduations in white enamel
  • Manufactured from borosilicate glass
  • Burettes and bottles should be ordered separately
Easy filling and automatic zero
  • Squeeze the polyethylene bottle to fill the burette
  • Release pressure to allow excess liquid to drain back into bottle and set the burette to zero
Leistungsmerkmale und Vorteile
  • Complies with ISO 385
  • Manufactured from borosilicate glass 3.3
  • With PTFE Rotaflo® Stopcock for grease-free performance and fine drop-wise control
  • 3mm bore PTFE key
  • Graduations in white enamel
  • Burettes and bottles should be ordered separately
  • Easy filling and automatic zero - Squeeze the polyethylene bottle to fill the burette - Release pressure to allow excess liquid to drain back into bottle and set the burette to zero
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Rotaflo is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)