Merck
SLW1000/11M
Pyrex® Griffin beakers, low form, with printed trace code
25 mL
Synonym(e):
Pyrex, SciLabware 1000/11M
Größe auswählen
10 EA
CHF 71.20
CHF 71.20
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 71.20
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
CHF 71.20
Check Cart for Availability
Materialien
borosilicate glass
white graduation
Leistungsmerkmale
low form
with printed trace code
Hersteller/Markenname
1000/11M
SciLabware
H × Durchm.
50 mm × 34 mm
Kapazität
25 mL
Allgemeine Beschreibung
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)