Merck
PL800GA-NIT-7
Plas-Labs replacement nitrile gloves
for 2 piece glove set, size 7 (for 8 in. ports)
Größe auswählen
1 PAIR
CHF 117.00
CHF 117.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 PAIR
CHF 117.00
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
CHF 117.00
Check Cart for Availability
Materialien
nitrile
Größe
7 (for 8 in. ports)
Leistungsmerkmale
reusable
for use with glove box sleeves
Verpackung
pack of 1 pair
Hersteller/Markenname
Plas-Labs 800GA-NIT-7
Allgemeine Beschreibung
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
Rechtliche Hinweise
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)