Merck
MT51343150
Mettler Toledo InLab® Viscous & Viscous Pro pH electrode
Without temperature probe
Größe auswählen
1 EA
CHF 650.00
CHF 650.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 650.00
About This Item
UNSPSC-Code:
41115604
NACRES:
NB.79
CHF 650.00
Check Cart for Availability
Materialien
glass membrane (polymer)
glass shaft (HA)
Beschreibung
S7 connector
Without cable
Hersteller/Markenname
Mettler-Toledo 51343150
L × B
40 mm × 6 mm
Allgemeine Beschreibung
The InLab® Viscous has been especially developed for viscous and highly glutinous samples.
The SteadyForce reference system guarantees the discharge of electrolytes, even with the most glutinous and fatty of samples, e.g. cosmetics, paints or resins.
The form has been chosen so that as little of the sample as possible adheres to the shaft and to facilitate cleaning. The result is a problem solver for most difficult sample types, the highly viscous materials.
  • high performance puncture electrode - especially designed for highly viscous
  • samples
  • easy to clean
  • SteadyForce reference system
Rechtliche Hinweise
InLab is a registered trademark of Mettler-Toledo, Inc.
SteadyForce is a trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)