Merck
MT51192107
Mettler Toledo LogStraight fingerprint reader
Größe auswählen
1 EA
CHF 510.00
CHF 510.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 510.00
About This Item
UNSPSC-Code:
41115600
NACRES:
NB.79
CHF 510.00
Check Cart for Availability
Hersteller/Markenname
Mettler-Toledo 51192107
Allgemeine Beschreibung
No more user names to enter, no more cumbersome password management. With LogStraight, the biometric fingerprint reader, the operators simply put their finger onto the reader and the DM density meter or RM refractometer knows immediately who they are.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)