Merck
MT11124300/UK
Mettler Toledo Printer RS-P25, RS232 interface
AC/DC input 230 V AC, UK plug
About This Item
UNSPSC-Code:
41115600
NACRES:
NB.61
Hersteller/Markenname
Mettler-Toledo 11124300/UK
Allgemeine Beschreibung
The RS-P25 Compact Printer provides simple printouts on lightfast or adhesive paper. It is a fast and reliable printer with a user friendly operation and a RS232 interface.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)