Merck
HAM201250
Hamilton® instrument syringe, for Boehringer Mannheim instruments, ES600
model, 1002, AccuDil (AD) end, volume 2.5 mL
About This Item
UNSPSC-Code:
42142600
NACRES:
NB.03
Materialien
Borosilicate glass/PTFE
Gastight syringe
Leistungsmerkmale
autoclavable: no
model 1002, AccuDil (AD) end
Hersteller/Markenname
Hamilton
Parameter
20-50 °C temp. range (68-122 °F)
200 psig pressure
Volumen
2.5 mL
Rechtliche Hinweise
Hamilton is a registered trademark of Hamilton Co.