GF13674595
Stainless steel - AISI 304l
rod, Fe/Cr18%/Ni 10%, 2.5 mm diameter, length 200 mm
Synonym(e):
FA147907
Alle Fotos(1)
About This Item
Form
rod
Hersteller/Markenname
Goodfellow 136-745-95
Länge
200 mm
Durchmesser
2.5 mm