Merck
GF11337735
Invar® - controlled expansion alloy
foil, Fe 64%/Ni 36%, thickness 0.5 mm, 8 mm diameter, temper annealed
Synonym(e):
FE020351
About This Item
UNSPSC-Code:
11101711
NACRES:
NA.23
Form
foil
Hersteller/Markenname
Goodfellow 113-377-35
Durchmesser
8 mm
8 mm
Dicke
0.5 mm
Verwandte Kategorien
Allgemeine Beschreibung
For updated SDS information please visit www.goodfellow.com
Rechtliche Hinweise
Product of Goodfellow
Invar is a registered trademark of Aperam Alloys Imphy