Merck
GF00367422
Palladium
foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%
Synonym(e):
Palladium, PD000270
About This Item
Lineare Formel:
Pd
CAS-Nummer:
Molekulargewicht:
106.42
MDL-Nummer:
UNSPSC-Code:
12141733
PubChem Substanz-ID:
NACRES:
NA.23
Assay
≥99.99%
Form
foil
Hersteller/Markenname
Goodfellow 003-674-22
Widerstandsfähigkeit
9.96 μΩ-cm, 20°C
L × Dicke
15 mm × 0.025 mm
bp
2970 °C (lit.)
mp (Schmelzpunkt)
1554 °C (lit.)
Dichte
12.02 g/cm3 (lit.)
SMILES String
[Pd]
InChI
1S/Pd
InChIKey
KDLHZDBZIXYQEI-UHFFFAOYSA-N
Allgemeine Beschreibung
For updated SDS information please visit www.goodfellow.com.
Rechtliche Hinweise
Product of Goodfellow
Lagerklassenschlüssel
13 - Non Combustible Solids
WGK
nwg
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Amanda J Hickman et al.
Nature, 484(7393), 177-185 (2012-04-14)
Copper and palladium catalysts are critically important in numerous commercial chemical processes. Improvements in the activity, selectivity and scope of these catalysts could drastically reduce the environmental impact, and increase the sustainability, of chemical reactions. One rapidly developing strategy for
Anton V Dubrovskiy et al.
Combinatorial chemistry & high throughput screening, 15(6), 451-472 (2012-01-26)
The iodocyclization of functionally-substituted alkynes provides an excellent way to prepare a wide range of iodoheterocycles, which can then be readily elaborated through palladium-catalyzed Suzuki-Miyaura, Sonogashira, Heck, Hartwig-Buchwald, and carbonylation processes into libraries of medicinally relevant heterocycles. The synthesis of
Update 1 of: Synthesis and functionalization of indoles through palladium-catalyzed reactions.
Sandro Cacchi et al.
Chemical reviews, 111(5), PR215-PR283 (2011-05-12)
Masahiro Toyota
Natural product communications, 8(7), 999-1004 (2013-08-29)
A novel palladium-catalyzed intramolecular oxidative alkylation of unactivated olefins is described. This protocol was devised to solve one of the drawbacks of the original palladium-catalyzed cycloalkenylation that we developed. We call this new procedure the 'second generation palladium-catalyzed cycloalkenylation'. This
Synthesis of heterocycles via palladium-catalyzed carbonylations.
Xiao-Feng Wu et al.
Chemical reviews, 113(1), 1-35 (2012-10-09)