GE63-0035-45
GE Exposure Cassette, Unmounted Screen
size 35 cm × 43 cm
Alle Fotos(1)
About This Item
UNSPSC-Code:
41105300
Verpackung
pkg of 1 ea
Hersteller/Markenname
Cytiva 63-0035-45
Verfügbarkeit
not available in Japan
Größe
35 cm × 43 cm
Allgemeine Beschreibung
The Exposure Cassette protects the screen from light during exposure to the sample.
For use with unmounted screens (not included).
Protects your screen from light during exposure.
For use with unmounted screens (not included).
Protects your screen from light during exposure.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)