Merck
CLS9985100
Pyrex® watch glasses
O.D. 100 mm
Synonym(e):
Corning® 9985
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Hersteller/Markenname
Corning 9985-100
AD
100 mm
Verwandte Kategorien
Allgemeine Beschreibung
Heavy-wall, borosilicate glass with fire-polished edge provides durability. May be slightly out-of-round due to O.D. variability.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)