Merck
CLS49501L
Pyrex® round-bottom vertical three neck distilling flask, with 24/40 joints
capacity 1,000 mL
Synonym(e):
bottom flask, flask round, flask round bottom, Corning® 4950
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.43
Materialien
borosilicate glass
round bottom flask
three-neck (Vertical)
Verbindung
joint ST/NS24/40
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard taper joint
Verpackung
pack of 1
case of 4
Hersteller/Markenname
Corning 4950-1L
Kapazität
1,000 mL
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.