Merck
CAD6330
1-way threaded end adapter (NPT)
MLL to 1/8-27 (NPT) thread (Plated Brass)
Größe auswählen
10 EA
CHF 401.00
CHF 401.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 401.00
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.03
CHF 401.00
Check Cart for Availability
Verpackung
pack of 10 ea
Hersteller/Markenname
Cadence Science 6330
Verwandte Kategorien
Allgemeine Beschreibung
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Rechtliche Hinweise
Micro-Mate is a registered trademark of Cadence Science
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1