BR94915
BRAND® standard ground joint funnel
funnel O.D. 80 mm
Synonym(e):
Funnels, Standard ground joint funnels
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Empfohlene Produkte
Materialien
polypropylene
Verbindung
joint ST/NS29/32
Verpackung
pack of 5 ea
Hersteller/Markenname
BRAND 94915
Trichter AD
80 mm
Allgemeine Beschreibung
Half-round body. Stem fits standard ground sockets. Ideal for filling liquids or powders into multi-neck flasks.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)