Merck
BR88670
BRAND® stopcock T-bore with nozzles
for tube I.D. 7 mm
Synonym(e):
PTFE, Stopcocks
Größe auswählen
1 EA
CHF 63.30
CHF 63.30
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 63.30
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
CHF 63.30
Check Cart for Availability
Materialien
PTFE
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 88670
ID
4 mm
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)