Merck
BR81939
BRAND® burette straight stopcock
for burette capacity 50 mL, PTFE key
Synonym(e):
Accessories, Burette stopcock, Burettes, Stopcocks
Größe auswählen
1 EA
CHF 51.20
CHF 51.20
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 51.20
About This Item
UNSPSC-Code:
41121808
NACRES:
NB.43
CHF 51.20
Check Cart for Availability
Materialien
PTFE key
amber
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 81939
Verwandte Kategorien
Allgemeine Beschreibung
BRAND burettes are always calibrated to deliver (TD, Ex).
Waiting times:
BLAUBRAND® - waiting time 30 sec. SILBERBRAND - no waiting time
A waiting time is usually not necessary if a titration is performed (see DIN EN ISO 385).
All BLAUBRAND® burettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)