Merck
BR701381
BRAND® Sealing Paddle
Größe auswählen
1 EA
CHF 9.80
CHF 9.80
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 9.80
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.13
CHF 9.80
Check Cart for Availability
Materialien
polyethylene
Sterilität
non-sterile
Hersteller/Markenname
BRAND 701381
Allgemeine Beschreibung
To securely and easily apply self-adhesive films. The sealing paddle helps to attach the film to the plate with a uniform pressure. Especially suited for plates with raised rim.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)