Merck
BR43320
BRAND® BLAUBRAND® density bottle, Gay-Lussac pattern, calibrated
capacity 25 mL, borosilicate glass
Synonym(e):
BLAUBRAND®, Gay-Lussac
Größe auswählen
1 EA
CHF 55.80
CHF 55.80
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 55.80
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.71
CHF 55.80
Check Cart for Availability
Materialien
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Verbindung
joint ST/NS neck10/19
Leistungsmerkmale
with cap
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 43320
Kapazität
25 mL
Allgemeine Beschreibung
DIN ISO 3507, type Gay-Lussac. Calibrated to contain (TC, In). Incl. individual certificate. Stopper NS 10/19 with capillary. Top of stopper ground and polished. The volume in cm³ is specified to a precision of 3 decimal places.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.