Merck
BR36982
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 500 mL, neck joint: ST/NS 14/23, glass stopper
Synonym(e):
BLAUBRAND®, USP, Volumetric flasks
Größe auswählen
2 EA
CHF 79.10
CHF 79.10
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
CHF 79.10
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
CHF 79.10
Check Cart for Availability
Materialien
borosilicate glass
glass stopper
Verbindung
joint: ST/NS neck14/23
Leistungsmerkmale
accuracy class A
Verpackung
pack of 2
Hersteller/Markenname
BRAND 36982
Kapazität
500 mL
Allgemeine Beschreibung
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)