Merck
BR152615
BRAND® tubing adapter
polypropylene, for tubing i.d., 6-10 to 9-13 mm
Synonym(e):
Tubing, Tubing adapters
Größe auswählen
10 EA
CHF 17.50
CHF 17.50
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 17.50
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
CHF 17.50
Check Cart for Availability
Materialien
polypropylene
Leistungsmerkmale
for tubing i.d. 6-10 to 9-13 mm
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 152615
Länge
59 mm
Allgemeine Beschreibung
The BRAND® tubing adapter is designed to provide a secure connection. Its translucent construction allows for easy visibility of fluid flow, facilitating monitoring during use.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)