Merck
BR150912
BRAND® watch glass
PTFE, diam. 80 mm
Synonym(e):
Dishes, PTFE, Watch glasses
Größe auswählen
1 EA
CHF 21.60
CHF 21.60
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 21.60
About This Item
UNSPSC-Code:
41121801
NACRES:
NB.43
CHF 21.60
Check Cart for Availability
Materialien
PTFE
Hersteller/Markenname
BRAND 150912
Durchmesser
80 mm
Allgemeine Beschreibung
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)