Merck
BMSW1000100
Sigma-Aldrich
Accuris F1 Grade Calibration Weight
100 g
Synonym(e):
milligram calibration weight, top pan balance weight, top pan calibration
Größe auswählen
1 EA
CHF 85.10
CHF 85.10
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 85.10
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
CHF 85.10
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific W1000-100
Allgemeine Beschreibung
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)