Merck
BMSV1002S8
Sigma-Aldrich
Aspire Laboratory Aspirator Accessories
Optional eight channel adapter, fine tip stainless steel
Synonym(e):
multichannel aspiration adapter, vacuum aspirator multichannel adapter
Größe auswählen
1 EA
CHF 164.00
CHF 164.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 164.00
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.13
CHF 164.00
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific V1002-S8
Verwandte Kategorien
Allgemeine Beschreibung
Accessories designed for use with Aspire laboratory aspirator.
The aspirator 8-channel fine capillary tip adapter is manufactured from stainless steel and enables fine control aspiration.
Rechtliche Hinweise
6538 is a trademark of American Type Culture Collection
Aspire is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)