Merck
BASMW1085
Sigma-Aldrich
Non-Aqueous Reference Electrode
Synonym(e):
Reference Electrode for Non-Aqueous Solutions
Größe auswählen
1 EACH
CHF 190.00
CHF 190.00
Check Cart for Availability
Bulk-Bestellung anfordern
Größe auswählen
Ansicht ändern
1 EACH
CHF 190.00
About This Item
UNSPSC-Code:
12352300
NACRES:
NA.23
CHF 190.00
Check Cart for Availability
Bulk-Bestellung anfordern
Materialien
PTFE cap
glass body
Beschreibung
frit tip with PTFE heat shrink
silver wire
Rechtliche Hinweise
Product of Bioanalytical Systems, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Questions
  1. Can I check the detailed specifications (thickness, usage environment, length, etc.) of this product?
    1 answer
    1. This is a product of Bioanalytical Systems, Inc. The length is 7.5 cm and the width at the tip of the probe is 5.7 mm. This electrode features a PTFE cap and glass body with a silver wire.
      Please see the supplier product image and drawing below.