Merck
BAF424002141
Scienceware® Lab Companion round vacuum desiccator
volume 10 L, amber polycarbonate (UV)
Größe auswählen
1 EA
CHF 595.00
CHF 595.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 595.00
About This Item
UNSPSC-Code:
41104510
NACRES:
NB.43
CHF 595.00
Check Cart for Availability
Materialien
amber polycarbonate (UV)
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F42400-2141
Volumen
10 L
10000 mL
Allgemeine Beschreibung
These high-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind. Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods. A Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange.
Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
Available in both cabinet and round styles, all models include a desiccant tray (desiccants sold separately).
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • Locking ring secures top and bottom halves when vacuum not in use; provides additional convenience and safety when moving the unit
  • Six models to choose from; 6, 10, and 20 L internal volume in clear or UV blocking amber
Rechtliche Hinweise
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1