Merck
BAF378400000
Sampling spatula, polypropylene
non-sterile
Größe auswählen
12 EA
CHF 37.60
CHF 37.60
Check Cart for Availability
Größe auswählen
Ansicht ändern
12 EA
CHF 37.60
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.35
CHF 37.60
Check Cart for Availability
Sterilität
non-sterile
Verpackung
pack of 12 ea
Hersteller/Markenname
Bel-Art F37840-0000
L × B × H
152 mm × 19 mm × 13 mm
Gesamt L
9 in.
Allgemeine Beschreibung
One-piece spatula is molded from polypropylene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.
  • V-shaped spatula is perfect for sampling granulated or semi solid material
  • Non-sterile
  • Use different spatula for different materials to reduce cross contamination
  • Sharp point will pierce most packaging
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)