Merck
934984
Sigma-Aldrich
Gold Microelectrode
diam. 10 μm
Synonym(e):
MF-2006
About This Item
UNSPSC-Code:
41111761
NACRES:
NA.21
Durchmesser
10 μm
Allgemeine Beschreibung
Microelectrodes consist of small diameter metal wires or fibers (5-100 µm) sealed within tempered glass bodies. We offer gold microelectrodes (Working electrodes) with 10 µm diameter for R&D applications in electrochemistry.Dimensions of the electrode are approximately 7 cm long and 4 mm OD. The electrical connection is a 0.060" gold-plated pin. Each microelectrode is visually inspected and electrochemically tested before shipping.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)