Merck
Pharmacological evaluation of NSAID-induced gastropathy as a "Translatable" model of referred visceral hypersensitivity.
World journal of gastroenterology (2017-10-04)
Michele Hummel, Terri Knappenberger, Meghan Reilly, Garth T Whiteside
RÉSUMÉ
To evaluate whether non-steroidal anti-inflammatory drugs (NSAIDs)-induced gastropathy is a clinically predictive model of referred visceral hypersensitivity. Gastric ulcer pain was induced by the oral administration of indomethacin to male, CD1 mice ( Results showed that two opioids and a GC-C agonist, morphine, asimadoline and linaclotide, respectively, the TRP antagonists, AMG9810 and HC-030031 and the sodium channel blocker, carbamazepine, elicited a dose- and/or time-dependent attenuation of referred visceral hypersensitivity, while the ASIC blocker, amiloride, was ineffective at all doses tested. Together, these findings implicate opioid receptors, GC-C, and sodium and TRP channel activation as possible mechanisms associated with visceral hypersensitivity. More importantly, these findings also validate NSAID-induced gastropathy as a sensitive and clinically predictive mouse model suitable for assessing novel molecules with potential pain-attenuating properties.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
AMG 9810, ≥98% (HPLC), powder