Merck
Purely coherent nonlinear optical response in solution dispersions of graphene sheets.
Nano letters (2011-11-19)
Rui Wu, Yingli Zhang, Shichao Yan, Fei Bian, Wenlong Wang, Xuedong Bai, Xinghua Lu, Jimin Zhao, Enge Wang
RÉSUMÉ
We have developed an efficient chemical exfoliation approach for the high-throughput synthesis of solution-processable, high-quality graphene sheets that are noncovalently functionalized by alkylamine. Purely coherent nonlinear optical response of these graphene sheets has been investigated, using near-infrared, visible, and ultraviolet continous wave and ultrafast laser beams. Spatial self-phase modulation has been unambiguously observed in the solution dispersions. Our results suggest that this coherent light scattering is due to a broadband, ultrafast, and remarkably huge third-order optical nonlinearity χ(3), which is a manifestation of the graphene's cone-shaped large-energy-scale band structure. Our experimental findings endow graphene new potentials in nonlinear optical applications.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Graphene, amine functionalized, avg. no. of layers, 1 ‑ 5