Merck
The place of tobramycin in lower respiratory tract infections (LRTI).
Journal of chemotherapy (Florence, Italy) (1995-08-01)
G Gialdroni Grassi
RÉSUMÉ
The Author provides a review of clinical experience with tobramycin as therapy for lower respiratory tract infections, in comparison to other aminoglycosides, including the pharmacokinetics and toxicity, dwelling on oto- and nephrotoxicity. The article includes a discussion of various dosing regimens of the aminoglycosides, focussing on efficacy and toxicity arising from once-daily administration. The Author then provides a more detailed description of tobramycin's pharmacokinetics, indications for its use, and the possibilities of once-daily dosing, concluding that toxicity is favorably influenced by a single daily administration as well as efficacy, and that patient compliance and reduced hospital costs are other advantages of this regimen.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tobramycine, Aminoglycoside antibiotic
Supelco
Tobramycine, Pharmaceutical Secondary Standard; Certified Reference Material
Tobramycine, European Pharmacopoeia (EP) Reference Standard
Tobramycine, European Pharmacopoeia (EP) Reference Standard