Merck
  • Uncatalyzed one-pot synthesis of highly substituted pyridazines and pyrazoline-spirooxindoles via domino SN/condensation/aza-ene addition cyclization reaction sequence.
Uncatalyzed one-pot synthesis of highly substituted pyridazines and pyrazoline-spirooxindoles via domino SN/condensation/aza-ene addition cyclization reaction sequence.
ACS combinatorial science (2013-04-30)
Nasrin Zohreh, Abdolali Alizadeh
RÉSUMÉ
A previously unknown class of highly substituted pyridazines and pyrazoline-spirooxinoles are easily prepared by an uncatalyzed one-pot three-component approach incorporating a domino SN/condensation/aza-ene addition cyclization reaction sequence. 1,1-Dihydrazino-2-nitroethylene (DHNE) which is generated in situ from the nucleophilic substitution (SN) reaction of hydrazine and 1,1-bis(methylthio)-2-nitroethylene (BMTNE), allowed to be condensed with active 1,2-dicarbonyl compounds followed by an intramolecular aza-ene addition cyclization to obtain the titled products depending on the type of 1,2-dicarbonyl. All reactions are easily performed and proceed with high efficiency under very simple and mild conditions without any catalyst and give good yields avoiding time-consuming, costly syntheses, and tedious workup and purification of products.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,1-Bis(methylthio)-2-nitroethylene, 95%